D-(+)-Galactose ≥99%

Supplier: Thermo Fisher Scientific

J15995-A7
101171-630EA 1762.06 USD
101171-630 101170-884 101172-510
D-(+)-Galactose ≥99%
D(+)-Galactose
Formula: C₆H₁₂O₆
Melting Pt: 164…168 °C
Density: 1.50
Storage Temperature: Ambient
MDL Number: MFCD00151230
CAS Number: 59-23-4
EINECS: 200-416-4
Merck Index: 14,4335
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&vid=3c02003b1d12c9bfd11c2e6faf5cc37d4325d208&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (HPLC) ≥99.0%
Identification (FTIR) Conforms
Residue on Ignition ≤0.5%
Loss on Drying ≤0.5%
Optical Rotation (c=4 in H₂O and NH₃) ≥77°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR