DNA sodium salt (Deoxyribonucleic acid sodium salt) (from E.coli)

Supplier: Thermo Fisher Scientific

Synonyms: Thymonucleic acid sodium salt, DNA sodium salt

J14380-MA
101171-590EA 198.14 USD
101171-590
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from E.coli)
DNA sodium salt (Deoxyribonucleic acid sodium salt)

E. coli genomic DNA is purified from E. coli type B cells, ATCC 11303 strain.


It has a single chromosome and the genome is 4,600,000 bp long. This genomic DNA is fragmented to some degree during purification yet it is characterized as a high molecular weight DNA.

Storage Temperature: Ambient
MDL Number: MFCD00130921
CAS Number: 9007-49-2

Order Now

Specification Test Results

Assay (unspecified), Hyperchromicity 0.27
Spectral Ratio, A260/A280 1.8

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR