Sodium dodecyl sulfate (SDS) solution 10%, Ultrapure
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
10% (w/v) solution of sodium dodecyl sulfate (SDS) in high purity dH2O.
- A(280 nm, 3%, 1 cm): ≤ 0.1
- A(230 nm, 3%, 1 cm): ≤ 0.2
Solution is 0.2 μm filtered and dispensed into durable, square Nalgene bottles.
Caution: Note: Precipitation of SDS is not unusual. Warm gently to redissolve.
Formula:
CH₃(CH₂)₁₁OSO₃Na Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 ADR: 4.1,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...