Formamide ≥99%, Ultrapure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Carbamaldehyde

J75828-A1
101171-418EA 229.25 USD
101171-418
Formamide ≥99%, Ultrapure
Formamide
Formula: HCONH₂
Boiling Pt: 210 °C
Melting Pt: 2 °C
Density: 1.134
Flash Pt: 154 °C(309 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 14,4237
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (unspecified) Absorbance (260 nm, 100%, 1 cm) ≤2.3
Assay (unspecified) Absorbance (280 nm, 100%, 1 cm) ≤0.15
Assay (unspecified) ≥99%
Conductivity ≤100 µmhos/cm
pH (50% v/v with H₂O) 6.0 to 8.0
Refractive Index (20 °C) 1.447 ±0.001
Copper (Cu) ≤0.2 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Zinc (Zn) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR