Glycine 98.5-101.5% (dry basis)

Supplier: Thermo Fisher Scientific

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

J16405-A1 J16405-C4 J16405-A7
101170-930EA 94.22 USD
101170-930 101170-932 101175-532
Glycine 98.5-101.5% (dry basis)
Glycine
Formula: NH₂CH₂COOH
Melting Pt: ∼ 245 °C (dec.)
Density: 1.595
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 14,4491
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Unspecified) 98.5 - 101.5%
Assay (Unspecified) - Hydrolysable Substances Passes Test
Chloride Content ≤70 ppm
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Impurity Content Sulfate (SO₄) ≤65 ppm
Loss on Drying ≤0.2%
Heavy Metals (Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR