EDTA disodium salt dihydrate 0.5 M in aqueous solution, Ultrapure for molecular biology
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate
0.5 M solution of EDTA.Na₂ in high purity dH₂O, pH 8.0.
Chelating agent used in many enzyme buffers and, at higher concentrations, as an enzyme inactivator.
Packaging: Dispensed into durable, square Nalgene bottles.
Formula:
C₁₀H₁₄N₂Na₂O₈·2H₂O MW: 372.24 g/mol Melting Pt: 255 Storage Temperature: Ambient |
MDL Number:
MFCD00150037 CAS Number: 6381-92-6 EINECS: 205-358-3 Merck Index: 13,03543 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...