Sodium dodecyl sulfate (SDS) ≥95% (dye basis)
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
J18220-36
J18220-A7
J18220-A1
101173-766EA
65
USD
101173-766
101170-512
101175-088
Sodium dodecyl sulfate (SDS) ≥95% (dye basis)
Sodium dodecyl sulphate (SDS)
Formula:
CH₃(CH₂)₁₁OSO₃Na Melting Pt: ∼ 206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 ADR: 4.1,II Merck Index: 14,8636 |
|
Specification Test Results
Assay (Dry Basis) | ≥95% |
Identification (FTIR) | Conforms |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...