Legato 100 Single Syringe Infuse Only Syringe Pump, KD Scientific
Supplier: KD Scientific
788100
10117-894EA 2667.36 USD
10117-894
Legato 100 Single Syringe Infuse Only Syringe Pump, KD Scientific
Pumps Syringe Pumps
Infuse only single syringe pump
  • Touch pad lock feature, LED light on front panel, USB port, I/O and TTL interface
  • Full metal chassis
  • Built in syringe table, RS485 interface
  • Advanced microstepping techniques
  • Continuous mode of operation
  • Spill dam
  • Programmable
Accomodates syringe sizes 0.5ul to 60 ml. User definable flow rates with selectable target volume or time values. High resolutuion touch screen display. Built in syringe table. Up to 30 lbs of linear force. USB control.
Certifications: UL, CSA, CUL, CE (Global), RoHS, WEEE.
Caution: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.