Legato 380 Emulsifier, KD Scientific
Supplier: KD Scientific
788380
10117-546EA 18995.87 USD
10117-546
Legato 380 Emulsifier, KD Scientific
Pumps Syringe Pumps
The Legato 380 emulsifying system utilizes a modified Legato 210P and Legato 270P
  • Custom emulsification program added to the software, only available with the Legato® 380 system
  • Selectable emulsifying rates and volumes
  • Adjustable duration
The pumps will automatically move the fluid back and forth between the syringes for a user determined volume, rate and time or number of cycles.Typical emulsifying rates are dependent on the size of syringe. The system accomodates syringe sizes 0.5ul to 140ml.
Certifications: UL, CSA, CUL, CE (Global), RoHS, WEEE.
Caution: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.