Legato 101 Dual Syringe Infuse Only Syringe Pump, KD Scientific
Supplier: KD Scientific
788101
10117-502EA 3851.57 USD
10117-502
Legato 101 Dual Syringe Infuse Only Syringe Pump, KD Scientific
Pumps Syringe Pumps
Infuse only dual syringe pump
  • Touch pad lock feature, LED light on front panel, USB port, I/O and TTL interface
  • Full metal chassis
  • Built in syringe table, RS485 interface
  • Advanced microstepping techniques
  • Continuous mode of operation
  • Spill dam
  • Programmable
Accomodates syringe sizes 0.5ul to 10ml. User definable flow rates with selectable target volume or time values. High resolutuion touch screen display. Built in syringe table. Up to 30 lbs of linear force. USB & RS232 control.
Certifications: UL, CSA, CUL, CE (Global), RoHS, WEEE.
Caution: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.