Legato 210 Dual Syringe Infuse/Withdraw Programmable Syringe Pump, KD Scientific
Supplier: KD Scientific
788212
10117-500EA 6201.26 USD
10117-500
Legato 210 Dual Syringe Infuse/Withdraw Programmable Syringe Pump, KD Scientific
Pumps Syringe Pumps
Infuse/Withdraw/Continuous dual syringe pump (syringes size 0.5ul to 140ml)
  • Touch pad lock feature, LED light on front panel, USB port, I/O and TTL interface
  • Full metal chassis
  • Built in syringe table, RS485 interface
  • Advanced microstepping techniques
  • Continuous mode of operation
  • Spill dam
  • Programmable
High resolutuion touch screen display. User definable flow rates with selectable target volume or time values. Programmable with 40 multi-step programs with constant rate, ramp, pulsed & stepped profiles.
Certifications: UL, CSA, CUL, CE (Global), RoHS, WEEE.
Caution: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.