Legato 130 Single Syringe Infuse/Withdraw Programmable Syringe Pump, KD Scientific
Supplier: KD Scientific
788130
10117-494EA 5036.84 USD
10117-494
Legato 130 Single Syringe Infuse/Withdraw Programmable Syringe Pump, KD Scientific
Pumps Syringe Pumps
Single syringe infuse/ withdraw with program option for 2 programs
  • Touch pad lock feature, LED light on front panel, USB port, I/O and TTL interface
  • Full metal chassis
  • Built in syringe table, RS485 interface
  • Advanced microstepping techniques
  • Continuous mode of operation
  • Spill dam
  • Programmable
50 steps each. Holds one syringes from 0.5µl to 1000µl. User definable flow rates with selectable target volume or time values to control the total infusion volume.
Certifications: UL, CSA, CUL, CE (Global), RoHS, WEEE.
Caution: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.