Stainless Steel PCR Workstation/UV Cabinet, Grant Instruments
Supplier: GRANT USA INC.
UVC/T-M-AR US
10117-478EA 5382 USD
10117-478 10117-480
Stainless Steel PCR Workstation/UV Cabinet, Grant Instruments
Fume Hoods and Enclosures PCR Workstation Enclosures
The UVC/T-M-AR is an efficient general purpose stainless steel PCR UV cabinet workstation for carrying out short-term procedures.
It is equipped with an open UV lamp installed in the upper part of the hood. UV-radiation from these lamps effectively disinfects the working area chamber, inactivating DNA / RNA fragments during 15-30 minutes of exposure. The digital timer controls the duration of the direct UV irradiation.
An additional daylight lamp provides a bright, clear working area. It is equipped with a high efficiency flow-type bactericidal UV cleaner-recirculator which provide constant decontamination inside the cabinet during operation.
The UV cleaner-recirculator consists of UV lamp, dust filters and a fan. The recirculation effect increases the maximum density of the UV light making it sufficiently effective for DNA / RNA inactivation. The recirculator processes 360 UV-cabinet volumes per hour, creating permanently aseptic conditions within the cabinet.
Certifications: CE marked.
Ordering information: The UVC/T-M-AR US does not have an internal socket.