Economy PCR Workstation/UV Cabinet, Grant Instruments
Supplier: GRANT USA INC.
UVT-B-AR USA
10117-476EA 5671.13 USD
10117-476
Economy PCR Workstation/UV Cabinet, Grant Instruments
Fume Hoods and Enclosures PCR Workstation Enclosures
The UVT-B-AR is an economical general purpose PCR UV cabinet in for carrying out short-term procedures
It is equipped with an open UV lamp installed in the upper part of the hood. UV-radiation from these lamps effectively disinfects the working area chamber, inactivating DNA / RNA fragments during 15-30 minutes of exposure. The UV light switches off automatically when door is opened.
The built-in UV cleaner-recirculator both increases the maximum density of UV light and generates 25 m³/h air flow exchange - prevents unwanted contamination and protects the user from direct UV light during manipulation.
The UV cleaner-recirculator consists of UV lamp, dust filters and a fan. The recirculation effect increases the maximum density of the UV light making it sufficiently effective for DNA / RNA inactivation. The recirculator processes 360 UV-cabinet volumes per hour, creating permanently aseptic conditions within the cabinet.
It is an economic model with stainless steel work surface, metal framework and shock proof glass panels.
Certifications: CE marked.
Ordering information: The UVT-B-AR US has one internal socket. The UVT-B-AR INLUS does not have an internal socket.