HPLC Safety Set, S.C.A.T.
307337
10117-458EA 1232.64 USD
10117-458
HPLC Safety Set, S.C.A.T.
This HPLC safety set can be used with any HPLC system
SafetyCap with air valve blocks solvent vapors, avoids underpressure, catches dust, keeps solvents clean and protects the user. SafetyWasteCap with exhaust filter blocks solvent vapors, avoids overpressure and protects the user. An indicator shows the filter status.
This set manages solvent supply and waste