Choline chloride ≥98.0% (dry basis)

Supplier: Thermo Fisher Scientific

Synonyms: 2-hydroxy-N,N,N-trimethylethanaminium chloride

J13410-P5 J13410-A9
101169-778EA 196 USD
101169-778 103728-066
Choline chloride ≥98.0% (dry basis)
Choline chloride
Formula: HOH₂CCH₂N(CH₃)₃Cl
MW: 139.62 g/mol
Melting Pt: ∼ 305 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011721
CAS Number: 67-48-1
EINECS: 200-655-4
Merck Index: 14,2206
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b015e76f45084bc46da3249578a85652cd6802&vid=85c8a0a1b152b861b3bc3ec8fe25a70509221d85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form White crystalline powder
Assay (unspecified): 1,4-Dioxane Passes test
Assay (unspecified) ≥98.0% (dry basis)
Comment Protect from moisture. This material is hygroscopic.
Identification (FTIR) Conforms
Residue on ignition ≤ 0.05%
Water Content (Karl Fischer Titration) ≤ 0.5%
Trace Metal: Heavy Metals (Pb) ≤ 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR