Loading Pipette Tips for DispoEquilibrium Dialyzer, Harvard Apparatus
Supplier: Harvard Apparatus
742222
101167-460PK 124.62 USD
101167-460
Loading Pipette Tips for DispoEquilibrium Dialyzer, Harvard Apparatus
Pipette Tips
The QuikPrep® DispoEquilibrium Dialyzer™ made of inert PTFE is suitable for binding and interaction studies. It allows maximum sample retention.This is useful for studies which use radiolabeled compounds.
  • Easy to use
  • Disposable
  • Small sample volumes: 25 to 75 μl each chamber
  • Rapid dialysis due to ultra-thin membrane
  • Leak-proof
High-quality regenerated cellulose membranes with MWCOs of 5000 and 10000 Daltons.