Anti-RNF2 Goat Polyclonal Antibody

Supplier: Prosci
46-304
10116-618EA 715.98 USD
10116-618
Anti-RNF2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 37-38 kDa band observed in lysates of cell lines of NIH3T3 and K562 (calculated MW of 37.7 kDa according to NP_009143.1). Recommended concentration: 0.3-1 ?g/mL.

RNF2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RNF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR