Anti-RAE1 Goat Polyclonal Antibody

Supplier: Prosci
46-264
10116-568EA 715.98 USD
10116-568
Anti-RAE1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 38-40 kDa band observed inHepG2 lysate (calculated MW of 41.0 kDa according to NP_003601.1 and NP_001015885.1). Recommended concentration: 0.03-0.1 ?g/mL. Immunohistochemistry: In paraffin embedded human brain cortex shows staining of neuronal cell bodies. Recommended concentration, 3-6 ?g/mL.

RAE1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RAE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR