Anti-PTBP2 Goat Polyclonal Antibody

Supplier: Prosci
46-244
10116-540EA 683.99 USD
10116-540
Anti-PTBP2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 35 kDa band in lysates of cell line NIH3T3 after 0.1 ?g/mL antibody staining.The 35 kDa band was successfully blocked by incubation with the immunizing peptide.

PTBP2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PTBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR