Large Stainless Steel Filter Holders, Advantec MFS
Supplier: Advantec MFS
302700 302800 302200 302100 351800 302500 302600 301900 302000 300800 300900
101105-008PK 8301.52 USD
101105-008 101105-150 101104-948 101104-602 101104-984 101105-148 101105-152 101105-054 101105-022 101105-722 101105-852
Large Stainless Steel Filter Holders, Advantec MFS
Filter Holders
Large Stainless Steel Pressure Filter Holders are offered in various sizes for batch type laboratory filtration using disc media.
Standard Holders are used in-line to filter gases and liquids such as culture media, solvents, photoresists. Sanitary Holders are threadless and can be disassembled for thorough cleaning. Fittings are 1.5" with tri-clamp. Use in-line to ultraclean or sterilize gases or liquids such as blood fractions, pure water and pharmaceutical applications.
The holder features a PTFE coated photoetched stainless steel filter support screen for autoclaving with filter in place. Unit comes standard with silicone o-ring. Other materials are available.
Units for filter diameters of 90 mm, 142 mm, and 293 mm are available in both Standard and Sanitary models. Optimal clean holder design minimizes resistance and maximizes flow rate.
Ordering information: Seal membrane by hand tightening locking wing nuts. Available in type 304 or 316 stainless steel. Advantec offers a complete family of large pressure holders and pressure vessels.