Pressure Filter Holders with Reservoir, Stainless Steel, Advantec MFS
Supplier: Advantec MFS
302300 361700 301500 361600 302400
101104-810PK 5252.05 USD
101104-810 101104-862 101105-212 101105-060 101105-046
Pressure Filter Holders with Reservoir, Stainless Steel, Advantec MFS
Filter Holders
These quality stainless steel holders are designed for filtering small batches of liquid without having to use a separate pressure vessel.
  • Suitable for hazardous toxicity testing and TCLP
  • Model KST47 has a photoetched 304 ss screen
  • KST90/142/293 screen is PTFE coated
  • Use for sterilizing, ultracleaning, and flow decay testing for pilot studies
There are separate ports for pressurizing the reservoir and for adding additional sample. A standard silicone o-ring seals the membrane internally by hand-tightening locking wing nuts. When using for intermittent fill operations, a back pressure support is recommended. Available in either type 304 or 316 stainless steel.
Ordering information: Replacement parts and alternate o-ring materials are available. KST293 models available. Assemblies can be PTFE coated by special order. Filters not included.