Blood Filter Strips, Advantec
Supplier: Advantec MFS
49010010
101104-522PK 21.23 USD
101104-522
Blood Filter Strips, Advantec
Filters Filter Paper Qualitative Filter Paper
These blood filter strips are used to track healthy and diseased animal populations safely, efficiently, and economically.
  • High purity cellulose fibers for consistent, uniform sampling
  • Each strip holds approximately 1 ml of blood or 0.04 ml serum
  • Filter strips require no refrigeration or pretreatment for sampling, storage, and analysis