MCE Membrane Filters, Advantec MFS
Supplier: Advantec MFS
A020A050A A045N047A A080V047A A045P047A A030A090C A100A142C A500A142C A080A050A A045A100C A100A090C A010A142C A045A142C A080X025B A080P025A A065A050A A065A047A A300A050A A500A050A A080N025A A020A293C A065A293C A500A293C A080X037A A080B050A A020K047A A080A142C A020A025A A045P025A A020A047A A030A047A A080A047A A100A047A A045V047A A080P047A A045V025A A045B047A A080B047A A045B037A A020A090C A300A090C A500A090C A045K047A A065A142C A300A142C A065B025A A045P137C A045A013A A080B037A A045A050A A045B013A A045B025A A100A293C A080A037A A045A037A A300A047A A045A102C A020A142C A030A142C A045A047A A080B013A A010A047A A045A293C A080A293C A020J047A A080N047A A080X037B A045J047A A010A050A A500A047A A045B050A A010A090C A500A037A A010A293C A300A293C A065A090C A080B025A A045U025A A045U047A A080U047A A010A013A A080A025A A080A090C A080X025A A080X047A A080X047B A100A013A A100A025A A300A013A A300A025A A500A013A A500A025A A045N013A A045N025A A045P013A A080N013A A010A025A A010A037A A020A013A A030A013A A030A025A A030A037A A030A293C A045A025A A045A090C A080A013A A020B025A A020B047A A065A013A A065A025A A065B047A A100B025A A100B047A A100A050A
101103-732PK 154.53 USD
101103-732 101103-808 101104-098 101104-120 101104-212 101104-264 101104-268 101103-740 101103-814 101104-216 101104-252 101104-258 101104-312 101104-366 101103-166 101103-736 101103-746 101103-750 101103-800 101103-948 101103-956 101103-966 101104-042 101104-182 101104-246 101104-262 101104-316 101104-364 101103-730 101103-734 101103-738 101103-742 101103-898 101104-122 101104-164 101104-176 101104-180 101104-194 101104-210 101104-220 101104-222 101104-250 101104-260 101104-266 101104-344 101103-578 101104-078 101104-196 101104-288 101104-340 101104-342 101103-962 101104-050 101104-048 101103-744 101103-816 101104-254 101104-256 101104-286 101104-346 101103-726 101103-952 101103-960 101104-060 101104-130 101104-146 101102-796 101103-728 101103-748 101104-178 101104-206 101103-932 101103-944 101103-964 101104-214 101104-348 101486-052 101486-054 101486-056 101486-058 101486-060 101486-062 101486-064 101486-066 101486-068 101486-070 101486-072 101486-074 101486-076 101486-080 101486-082 101486-086 101486-088 101486-090 101486-092 101486-094 101486-096 101486-098 101486-100 101486-102 101486-104 101486-106 101486-108 101486-112 101486-114 101486-116 101486-118 101486-120 101486-122 101486-124 101486-132 102407-620 102407-622 102407-624 102407-626 102407-628 102407-632 102407-634 102407-636 102407-638 102407-640
MCE Membrane Filters, Advantec MFS
Filters Membrane Filters
Mixed Cellulose Esters (MCE) membrane is a general purpose filter used in many applications for microbiological analysis and air monitoring. Contrasting grid lines are tested to assure freedom from grid line inhibition and facilitate colony/particle counting. Black and Green membranes provide a contrastng background for counting light colored particulates and help to minimize eye fatique.
  • Available pore sizes include 0.10, 0.20, 0.30, 0.45, 0.65, 0.80, 1.0, 3.0 and 5.0 µm
  • Available membrane colors include white, black and green
  • Surfaces are either gridded or plain with or without a hydrophobic edge (HE)
MCE membranes are manufactured to comply with the ASTM and the Standard Methods for the Examination of Water and Wastewater. MCE 0.80 µm filters meet NIOSH requirements for air sampling applications such as metals analysis and aesbestos monitoring. Quality control tests are performed on each lot and QA Certificates are available.
Advantec Opticlear MCE offers superior optical clarity to view collected particles. Clears completely, providing an optimal surface for accurate particle identification. Cutting and packaging are performed under cleanroom conditions to minimize contamination.
Packaging: Membrane filters are bulk packed and non sterile.