Swine Serum, Vector Laboratories
Supplier: Vector Laboratories
S-4000-20
101098-388EA 199.38 USD
101098-388
Swine Serum, Vector Laboratories
Sera (Serum)
Normal Swine serum can be used as a blocking reagent in Immunoassays.
  • Pooled sample
  • Heat-treated and centrifuged
  • Tested for possible cross-reactives
It is a pooled sample collected from healthy adult animals. The serum is heat-treated and centrifuged to remove precipitates and then filtered. Each serum is tested with the appropriate antibody to check for possible cross-reactivities. The sera are supplied undiluted with 0.08% sodium azide as a preservative.