Antigen Unmasking Solution, Vector Laboratories

Supplier: Vector Laboratories

H-3300-250
101098-054EA 252 USD
101098-054
Antigen Unmasking Solution, Vector Laboratories
Protein Expression Protein Expression Reagents

This antigen unmasking solutions are highly effective at revealing antigens in formalin-fixed, paraffin embedded tissue sections when used in combination with a high temperature treatment procedure


Applications: Immunohistochemistry, Immunocytochemistry and Immunofluorescence.


This citrate-based solution have a pH of 6.0.


Delivery information: Supplied as convenient 100X concentrated stock and sufficient for preparation of 25 L of working solution.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR