4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222)

Supplier: USP
Warning

1272918
101097-388EA 1523.08 USD
101097-388
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222)
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222)
Formula: C₁₈H₃₆N₂O₆
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00005111
CAS Number: 23978-09-8
EINECS: 245-962-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a74ac532561e4fecf111f3260b402bcd1fb0af9&vid=13491901da172b5971e6d7f330c893460a77cd23&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR