Particle count set (2 blanks and 2 suspensions) USP
Supplier: USP
1500502
101097-190EA 1464.62 USD
101097-190
Particle count set (2 blanks and 2 suspensions) USP
Standards Particle Size Standards
Particle count - Suspension - 25 mL: 15 µm Polystyrene microspheres in aqueous suspension
Particle count - Blank - 25 mL: Contains the aqueous phase without added particle