Cyanoguanidine

Supplier: USP

1396310
101097-130EA 1289.23 USD
101097-130
Cyanoguanidine
Cyanoguanidine
Formula: (H₂N)₂CNCN
Boiling Pt: 143 °C (1013 hPa)
Density: 1.404 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008066
CAS Number: 461-58-5
EINECS: 207-312-8
Merck Index: 13,03119

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR