Isoflurane Related Compound B USP Reference Standard

Supplier: USP
Warning

1349025
101097-074EA 1460 USD
101097-074
Isoflurane Related Compound B USP Reference Standard
Difluoromethyl-2,2,2-trifluoroethyl ether
Formula: C₃H₃F₅O
MDL Number: MFCD00153127
CAS Number: 1885-48-9

Order Now

Specification Test Results

Molecular formula C3H3F5O
Molecular weight 150.05
Synonym 2,2,2-Trifluoroethyl difluoromethyl ether


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR