2-Amino-4-chlorophenol

Supplier: USP
Warning

Synonyms: 5-Chloro-2-hydroxyaniline

1130527
101096-878EA 1389.23 USD
101096-878
2-Amino-4-chlorophenol
2-Amino-4-chlorophenol
Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 140…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00007694
CAS Number: 95-85-2
EINECS: 202-458-9
UN: 2673
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR