3-Aminophenol ≥99%

Supplier: USP
Warning

Synonyms: 3-Hydroxyaniline, m-Aminophenol

1026004
101096-780EA 1289.23 USD
101096-780
3-Aminophenol ≥99%
3-Aminophenol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 122 °C
Density: 1.275 g/cm³ (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00007786
CAS Number: 591-27-5
EINECS: 209-711-2
UN: 2512
ADR: 6.1,III
Merck Index: 13,00459

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR