Dihydrocapsaicin

Supplier: USP

Synonyms: N-(4-Hydroxy-3-methoxybenzyl)-8-methylnonanamide, 8-Methyl-N-vanillylnonanamid

1200600
101096-704EA 696.92 USD
101096-704
Dihydrocapsaicin
Dihydrocapsaicin
Formula: C₁₈H₂₉NO₃
MW: 307.43
MDL Number: MFCD00057794
CAS Number: 19408-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR