Cholecalciferol (Vitamin D3)

Supplier: USP

1131009
101096-454EA 452.31 USD
101096-454
Cholecalciferol (Vitamin D3)
Cholecalciferol (Vitamin D3)
Formula: C₂₇H₄₄O
MDL Number: MFCD00078131
CAS Number: 67-97-0
EINECS: 200-673-2
UN: 2811
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR