Polyethylene terephthalate

Supplier: USP

1546900
101095-858EA 452.31 USD
101095-858
Polyethylene terephthalate
Polyethylene terephthalate

  • 3 Strips 2'' × 2 1/2''

Formula: (C₁₀H₈O₄)n
MDL Number: MFCD00084422
CAS Number: 25038-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR