2-Phenoxyethanol

Supplier: USP

1526200
101095-788EA 469.23 USD
101095-788
2-Phenoxyethanol
2-Phenoxyethanol
Formula: C₆H₅OCH₂CH₂OH
MDL Number: MFCD00002857
CAS Number: 122-99-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27cc4d39b806528798164961c23a1285d41d6a9&vid=a46b721cc69101d0c50dc49d9687445b395a3766&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR