Mupirocin USP Reference Standard

Supplier: USP
Warning

Synonyms: 9-(((E)-4-((2S,3R,4R,5S)-3,4-Dihydroxy-5-(((2S,3S)-3-((2S,3S)-3-hydroxybutan-2-yl)oxiran-2-yl)methyl)tetrahydro-2H-pyran-2-yl)-3-methylbut-2-enoyl)oxy)nonanoic acid

1448901
101095-584EA 435.38 USD
101095-584
Mupirocin USP Reference Standard
Mupirocin
Formula: C₂₆H₄₄O₉
MW: 500.629 g/mol
MDL Number: MFCD01711620
CAS Number: 12650-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR