D-(-)-Mannitol

Supplier: USP

1375105
101095-386EA 489.23 USD
101095-386
D-(-)-Mannitol
D(-)-Mannitol
Formula: HOH₂C(CH(OH))₄CH₂OH
Boiling Pt: 290…295 °C (4 hPa)
Melting Pt: 164…169 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR