Manganese(II) sulfate monohydrate

Supplier: USP
Warning

Synonyms: Manganese sulphate monohydrate, Manganese sulfate monohydrate, Manganous sulfate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate, Manganese(II) sulfate hydrate

1375149
101095-384EA 372.31 USD
101095-384
Manganese(II) sulfate monohydrate
Manganese (II) sulphate monohydrate
Formula: MnSO₄·1H₂O
MW: 169.02 g/mol
Melting Pt: 700
Density: 2.95 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9 III
Merck Index: 13,05763

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR