Manganese(II) chloride tetrahydrate analytical standard

Supplier: USP
Warning

1375127
101095-382EA 387.69 USD
101095-382
Manganese(II) chloride tetrahydrate analytical standard
Manganese (II) chloride tetrahydrate
Formula: MnCl₂·4H₂O
Boiling Pt: 1190 °C (1013 hPa)
Melting Pt: 58…59 °C
Density: 2.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149792
CAS Number: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9,III
Merck Index: 13,05751

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR