Magnesium carbonate hydrate

Supplier: USP

1374226
101095-358EA 393.85 USD
101095-358
Magnesium carbonate hydrate
Magnesium carbonate hydrate
Formula: MgCO₃·xH₂O
Density: 3.03 g/cm³ (16 °C)
MDL Number: MFCD00151022
CAS Number: 23389-33-5
EINECS: 208-915-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR