Lanolin stabilised

Supplier: USP

Synonyms: Wool fat

1356880
101095-306EA 416.92 USD
101095-306
Lanolin stabilised
Lanolin
Melting Pt: 38…44 °C
Density: 0.932…0.945 g/cm³ (15 °C)
Flash Pt: 113 °C (closed cup)
MDL Number: MFCD00081740
CAS Number: 8006-54-0
EINECS: 232-348-6
Merck Index: 12,05371

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR