Lactose

Supplier: USP

1356676
101095-296EA 513.85 USD
101095-296
Lactose
Lactose
Formula: C₁₂H₂₂O₁₁
Boiling Pt: 592 °C (1013 hPa)
Melting Pt: 222.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00151251
CAS Number: 63-42-3
EINECS: 200-559-2
Merck Index: 12,05356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR