Isoflurane USP Reference Standard

Supplier: USP
Warning

Synonyms: 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether

1349003
101095-254EA 403.08 USD
101095-254
Isoflurane USP Reference Standard
Isoflurane
Formula: C₃H₂ClF₅O
MW: 184.49 g/mol
Boiling Pt: 47…50 °C
MDL Number: MFCD00066609
CAS Number: 26675-46-7
EINECS: 247-897-7
UN: 3334
ADR: 9 III

Order Now

Specification Test Results

Molecular formula C3H2ClF5O
Molecular weight 184.49
Volatile 100 %
Specific gravity 1.45 - 1.496
Synonym 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR