Hydroquinone

Supplier: USP
Danger

1324002
101095-166EA 420 USD
101095-166
Hydroquinone
Hydroquinone
Formula: C₆H₄(OH)₂
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 172 °C
Density: 1.332 g/cm³ (20 °C)
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00002339
CAS Number: 123-31-9
EINECS: 204-617-8
UN: 3077
ADR: 9,III
Merck Index: 13,04833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=203cfb319003b2b4489cbe3ab15876fc37f27cb8&vid=6d76260c677491e292c03637f8a08e9194282d48&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR