(±)-2-Methyl-2,4-pentanediol

Supplier: USP
Warning

1308200
101095-128EA 453.85 USD
101095-128
(±)-2-Methyl-2,4-pentanediol
(±)-2-Methyl-2,4-pentanediol
Formula: C₆H₁₄O₂
Boiling Pt: 198 °C (1013 hPa)
Melting Pt: –40 °C
Density: 0.923 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00004547
CAS Number: 107-41-5
EINECS: 203-489-0
Merck Index: 13,04730

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR