Guaifenesin

Supplier: USP

1301007
101095-104EA 435.38 USD
101095-104
Guaifenesin
Guaifenesin
Formula: C₁₀H₁₄O₄
MDL Number: MFCD00016873
CAS Number: 93-14-1
EINECS: 202-222-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR