Gemfibrozil

Supplier: USP

1288500
101095-072EA 403.08 USD
101095-072
Gemfibrozil
Gemfibrozil
Formula: C₁₅H₂₂O₃
Boiling Pt: 395 °C (1013 hPa)
Melting Pt: 59…61 °C
MDL Number: MFCD00079335
CAS Number: 25812-30-0
EINECS: 247-280-2
Merck Index: 12,04394

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR