Fumaric acid

Supplier: USP
Warning

1286708
101095-054EA 489.23 USD
101095-054
Fumaric acid
Fumaric acid
Formula: C₄H₄O₄
Boiling Pt: 356 °C (1013 hPa)
Melting Pt: 287 °C
Density: 1.625 g/cm³ (20 °C)
Flash Pt: 273 °C
MDL Number: MFCD00002700
CAS Number: 110-17-8
EINECS: 203-743-0
Merck Index: 13,04308

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR