5-Fluorouracil

Supplier: USP
Danger

Synonyms: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU

1279000
101095-036EA 435.38 USD
101095-036
5-Fluorouracil
5-Fluorouracil
Formula: C₄H₃FN₂O₂
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 282…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00006018
CAS Number: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1,III
Merck Index: 13,04208

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR